3,4-Dichloro-6-nitrotoluene ≥98.0% (by GC)

供应商: TCI

Synonyms: 4,5-Dichloro-2-nitrotoluene, 1,2-Dichloro-4-methyl-5-nitrobenzene

D3271-25G D3271-5G
TCIAD3271-25GEA 0 CNY
TCIAD3271-25G TCIAD3271-5G
3,4-Dichloro-6-nitrotoluene ≥98.0% (by GC)
3,4-Dichloro-6-nitrotoluene
分子式: C₇H₅Cl₂NO₂
分子量: 206.03 g/mol
MDL编号: MFCD01109847
CAS号: 7494-45-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42f93a2f4036e905dddfa169d95eede0ab04de6e&vid=42f93a2f4036e905dddfa169d95eede0ab04de6e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 60.0 to 64.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR