4-溴-2-氟-1-硝基苯 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 4-Bromo-2-fluoronitrobenzene

B3062-25G B3062-5G
TCIAB3062-25GEA 0 CNY
TCIAB3062-25G TCIAB3062-5G
4-溴-2-氟-1-硝基苯 ≥98.0% (by GC)
4-溴-2-氟-1-硝基苯
分子式: C₆H₃BrFNO₂
分子量: 220 g/mol
MDL编号: MFCD01930221
CAS号: 321-23-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf57a070d48f6791b4e21fdb8a1cefa32eb1897d&vid=bf57a070d48f6791b4e21fdb8a1cefa32eb1897d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 84.0 to 90.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR