o,o,o-Tritolyl phosphate ≥97.0% (通过GC)

供应商: TCI
Danger

P1331-25G P1331-5G
TCIAP1331-25GEA 0 CNY
TCIAP1331-25G TCIAP1331-5G
o,o,o-Tritolyl phosphate ≥97.0% (通过GC)
o,o,o-Tritolyl phosphate
分子式: C₂₁H₂₁O₄P
沸点: 410°C
溶点: 11 °C
密度: 1,1955 g/cm³ (20 °C)
闪点: 225°C
MDL编号: MFCD00041906
CAS号: 78-30-8
EINECS: 201-103-5
UN号: 2574
ADR: 6.1,II
Merck Index: 14,09764

现在订购

产品技术规格

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR