5-溴-2-噻吩硼酸 (contains varying amounts of Anhydride)

供应商: TCI
Warning

Synonyms: 5-溴噻吩-2-硼酸

B2862-1G B2862-5G
TCIAB2862-1GEA 0 CNY
TCIAB2862-1G TCIAB2862-5G
5-溴-2-噻吩硼酸 (contains varying amounts of Anhydride)
5-溴-2-噻吩硼酸
分子式: C₄H₄BBrO₂S
分子量: 206.85 g/mol
存储条件: Refrigerator
MDL编号: MFCD02094028
CAS号: 162607-17-2

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR