3,5-双(三氟甲基)苯硼酸 (contains varying amounts of Anhydride)

供应商: TCI
Warning

Synonyms: 3,5-Bis(trifluoromethyl)benzeneboronic acid, (3,5-Bis(trifluoromethyl)phenyl)metaboric acid, (3,5-Bis(trifluoromethyl)phenyl)boronic acid, Dihydroxy(3,5-bis(trifluoromethyl)phenyl)borane

B1886-1G B1886-5G
TCIAB1886-1GEA 0 CNY
TCIAB1886-1G TCIAB1886-5G
3,5-双(三氟甲基)苯硼酸 (contains varying amounts of Anhydride)
3,5-双(三氟甲基)苯硼酸
分子式: C₈H₅BF₆O₂
分子量: 257.93 g/mol
MDL编号: MFCD00051850
CAS号: 73852-19-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ed5a24fbf4a55ac4cfc505fc9827ad159314dac&vid=9ed5a24fbf4a55ac4cfc505fc9827ad159314dac&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) 95.0 to 108.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR