邻甲基苯硼酸 (contains varying amounts of Anhydride)

供应商: TCI
Warning

Synonyms: 2-甲基苯硼酸

M1313-25G M1313-5G
TCIAM1313-25GEA 0 CNY
TCIAM1313-25G TCIAM1313-5G
邻甲基苯硼酸 (contains varying amounts of Anhydride)
邻甲基苯硼酸
分子式: C₇H₉BO₂
分子量: 135.96 g/mol
MDL编号: MFCD00093526
CAS号: 16419-60-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a1ba1bd97380d6b75038543068db437524f5301&vid=3a1ba1bd97380d6b75038543068db437524f5301&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) 98.0 to 116.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR