3-(2-羟乙基)吲哚 ≥98.0% (by GC)
供应商: TCI
Synonyms:
色醇, 3-吲哚乙醇
I0030-1G
I0030-25G
I0030-5G
TCIAI0030-1GEA
0
CNY
TCIAI0030-1G
TCIAI0030-25G
TCIAI0030-5G
3-(2-羟乙基)吲哚 ≥98.0% (by GC)
3-(2-羟乙基)吲哚
分子式:
C₁₀H₁₁NO 分子量: 161.2 g/mol 沸点: 179°C |
MDL编号:
MFCD00005659 CAS号: 526-55-6 EINECS: 208-393-2 Merck Index: 14,09798 |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 57.0 to 60.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...