异戊二烯 ≥99.0% (by GC) stabilised
供应商: TCI
|
Danger
|
Synonyms:
2-甲基-1,3-丁二烯
Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC)
I0160-25ML
I0160-500ML
TCIAI0160-25MLEA
0
CNY
TCIAI0160-25ML
TCIAI0160-500ML
异戊二烯 ≥99.0% (by GC) stabilised
异戊二烯
分子式:
C₅H₈ 沸点: 35°C 溶点: -146°C 密度: 0,681 g/cm³ (20 °C) 闪点: -54°C 存储条件: Refrigerator |
MDL编号:
MFCD00008600 CAS号: 78-79-5 EINECS: 201-143-3 UN号: 1218 ADR: 3,I Merck Index: 14,05201 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...