2-溴苯乙醇 ≥98.0% (by GC)

供应商: TCI

B3516-25G B3516-5G
TCIAB3516-25GEA 0 CNY
TCIAB3516-25G TCIAB3516-5G
2-溴苯乙醇 ≥98.0% (by GC)
2-溴苯乙醇
分子式: C₈H₉BrO
分子量: 201.06 g/mol
沸点: 132°C
MDL编号: MFCD00093566
CAS号: 1074-16-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf9f7718c4b08de68808441e15fab1967641a23b&vid=cf9f7718c4b08de68808441e15fab1967641a23b&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR