2,3,4-三氟-6-硝基苯胺 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2-Nitro-4,5,6-trifluoroaniline, 2,3,4-Trifluoro-6-nitroaniline
T2783-1G
T2783-5G
TCIAT2783-1GEA
0
CNY
TCIAT2783-1G
TCIAT2783-5G
2,3,4-三氟-6-硝基苯胺 ≥98.0% (by GC)
2,3,4-三氟-6-硝基苯胺
分子式:
C₆H₃F₃N₂O₂ |
MDL编号:
MFCD00174089 CAS号: 148416-38-0 |
|
产品技术规格
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 58.0 to 62.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...