1-苯基-1,3-丁二酮 ≥98.0% (by GC)
供应商: TCI
Synonyms:
苯甲酰丙酮
P0160-25G
P0160-500G
TCIAP0160-25GEA
0
CNY
TCIAP0160-25G
TCIAP0160-500G
1-苯基-1,3-丁二酮 ≥98.0% (by GC)
1-苯基-1,3-丁二酮
分子式:
C₆H₅C(O)CH₂C(O)CH₃ 沸点: 262°C 溶点: 57…59 °C 密度: 1,0899 g/cm³ (60 °C) |
MDL编号:
MFCD00008786 CAS号: 93-91-4 EINECS: 202-286-4 |
|
产品技术规格
Appearance | White to Light yellow powder to crystaline |
Purity (GC) | min. 98.0 % |
Melting point | 57.0 to 60.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...