MOPS钠盐 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 3-(N-吗啉基)丙磺酸, 4-吗啉丙磺酸

M0755-25G M0755-250G
TCIAM0755-25GEA 0 CNY
TCIAM0755-25G TCIAM0755-250G
MOPS钠盐 ≥98.0% (by titrimetric analysis)
MOPS钠盐
分子式: C₇H₁₄NNaO₄S
分子量: 231.25 g/mol
存储条件: Ambient
MDL编号: MFCD00064350
CAS号: 71119-22-7
EINECS: 428-420-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1e60c1fa2f5cbfa2202e7039231ce310eade30e&vid=b1e60c1fa2f5cbfa2202e7039231ce310eade30e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR