(S)-(+)-1,2-异亚丙基甘油 ≥98.0% (by GC)

供应商: TCI

Synonyms: D-1,2-O-isopropylidene-sn-glycerol, (S)-(+)-2,2-二甲基-1,3-二氧戊环-4-甲醇, D-(+)-丙酮缩甘油, 1,2-Isopropylidene-sn-glycerol, D-α,β-异亚丙基甘油

D1691-1G D1691-25G D1691-5G
TCIAD1691-1GEA 0 CNY
TCIAD1691-1G TCIAD1691-25G TCIAD1691-5G
(S)-(+)-1,2-异亚丙基甘油 ≥98.0% (by GC)
(S)-(+)-1,2-异亚丙基甘油
分子式: C₆H₁₂O₃
沸点: 90°C
密度: 1,0704 g/cm³ (20 °C)
闪点: 80°C
存储条件: Refrigerator
MDL编号: MFCD00063239
CAS号: 22323-82-6
EINECS: 244-910-8
Merck Index: 14,05213
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40d669c8829cf90ec8d4024aa7ed24d5cdc79a61&vid=40d669c8829cf90ec8d4024aa7ed24d5cdc79a61&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 98.0 ee%
Specific rotation [a]20/D +13.5 to +14.5 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR