3-硝基联苯 ≥99.0% (by GC)
供应商: TCI
Warning
Synonyms: 3-Nitro-1,1'-biphenyl
N0193-1G N0193-25G
TCIAN0193-1GEA 0 CNY
TCIAN0193-1G TCIAN0193-25G
3-硝基联苯 ≥99.0% (by GC)
3-硝基联苯
分子式: C₁₂H₉NO₂
沸点: 204°C
溶点: 62 °C
密度: 1,305 g/cm³ (20 °C)
MDL编号: MFCD00007245
CAS号: 2113-58-8
EINECS: 218-305-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcc771532b8975dc66c7b68386b44c9d5d7017dd&vid=dcc771532b8975dc66c7b68386b44c9d5d7017dd&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 99.0 %
Melting point 58.0 to 61.0 deg-C