3-叔丁基-4-羟基苯甲醚 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
3-BHA, BHA, 2-叔丁基-4-甲氧基苯酚
B0723-25G
B0723-500G
TCIAB0723-25GEA
0
CNY
TCIAB0723-25G
TCIAB0723-500G
3-叔丁基-4-羟基苯甲醚 ≥98.0% (by GC)
3-叔丁基-4-羟基苯甲醚
分子式:
C₁₁H₁₆O₂ 分子量: 180.25 g/mol |
MDL编号:
MFCD00040484 CAS号: 121-00-6 EINECS: 204-442-7 UN号: 3077 ADR: 9 III |
|
产品技术规格
Appearance | White to Light yellow powder to crystaline |
Purity (GC) | min. 98.0 % |
Melting point | 59.0 to 65.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...