2-溴-4′-甲氧基苯乙酮 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: 4-甲氧基苯甲酰基甲基溴, ω-溴-4-甲氧基苯乙酮

M1004-25G M1004-5G
TCIAM1004-25GEA 0 CNY
TCIAM1004-25G TCIAM1004-5G
2-溴-4′-甲氧基苯乙酮 ≥98.0% (by GC)
2-溴-4′-甲氧基苯乙酮
分子式: C₉H₉BrO₂
分子量: 229.07 g/mol
沸点: 285
存储条件: Refrigerator
MDL编号: MFCD00000201
CAS号: 2632-13-5
EINECS: 220-118-8
UN号: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7a0ab86141ca9c7e80b496894bcfbc71e025792&vid=b7a0ab86141ca9c7e80b496894bcfbc71e025792&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 69.0 to 72.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR