1-Bromo-4-heptylbenzene 96,0 GC_ASSAY_METHOD Stabilised
供应商: TCI
Synonyms:
(4-Bromophenyl)heptane
Stabilization:
Copper chip_stab
B1608-1G
B1608-5G
TCIAB1608-1GEA
0
CNY
TCIAB1608-1G
TCIAB1608-5G
1-Bromo-4-heptylbenzene 96,0 GC_ASSAY_METHOD Stabilised
1-Bromo-4-heptylbenzene
分子式:
C₁₃H₁₉Br 分子量: 255.2 g/mol 沸点: 190°C |
MDL编号:
MFCD00061115 CAS号: 76287-49-5 |
|
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...