水杨酰肼 ≥98.0% (by HPLC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
水杨酸酰肼, 2-羟基苯甲酰肼
S0242-25G
S0242-500G
TCIAS0242-25GEA
0
CNY
TCIAS0242-25G
TCIAS0242-500G
水杨酰肼 ≥98.0% (by HPLC, titration analysis)
水杨酰肼
分子式:
C₇H₈N₂O₂ 分子量: 152.15 g/mol |
MDL编号:
MFCD00007599 CAS号: 936-02-7 EINECS: 213-311-3 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 148.0 to 152.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...