正十二烷基硫醚 ≥96.0% (by GC)

供应商: TCI
Warning

Synonyms: Dodecyl sulphide

D1327-25G D1327-500G
TCIAD1327-25GEA 0 CNY
TCIAD1327-25G TCIAD1327-500G
正十二烷基硫醚 ≥96.0% (by GC)
正十二烷基硫醚
分子式: C₂₄H₅₀S
分子量: 370.73 g/mol
沸点: 465
溶点: 40
MDL编号: MFCD00008975
CAS号: 2469-45-6
EINECS: 219-587-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=323a573a30d090641d3bc02804e02da5cedb745c&vid=323a573a30d090641d3bc02804e02da5cedb745c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 41.0 to 43.0 deg-C
Solubility in Toluene very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR