4-甲氧基苯硼酸 (contains varying amounts of Anhydride)

供应商: TCI
Warning

Synonyms: 对甲氧基苯硼酸

M1252-25G M1252-5G
TCIAM1252-25GEA 0 CNY
TCIAM1252-25G TCIAM1252-5G
4-甲氧基苯硼酸 (contains varying amounts of Anhydride)
4-甲氧基苯硼酸
分子式: C₇H₉BO₃
分子量: 151.96 g/mol
存储条件: Ambient
MDL编号: MFCD00039139
CAS号: 5720-07-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71d7d36b7608726cd802fea10658e3e682001667&vid=71d7d36b7608726cd802fea10658e3e682001667&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) 97.0 to 113.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR