1,3-丙酮二羧酸 ≥95.0% (by titrimetric analysis)
供应商: TCI
|
Warning
|
Synonyms:
3-氧代戊二酸
K0006-25G
K0006-250G
TCIAK0006-25GEA
0
CNY
TCIAK0006-25G
TCIAK0006-250G
1,3-丙酮二羧酸 ≥95.0% (by titrimetric analysis)
1,3-丙酮二羧酸
分子式:
C₅H₆O₅ 沸点: 323 °C (1013 hPa) 溶点: 138 °C 密度: 1,55 g/cm³ (20 °C) 存储条件: Freezer |
MDL编号:
MFCD00002711 CAS号: 542-05-2 EINECS: 208-797-9 Merck Index: 14,00068 |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...