5-氯-2-氟吡啶 ≥96.0% (by GC)

供应商: TCI
Danger

C2303-25G C2303-5G
TCIAC2303-25GEA 0 CNY
TCIAC2303-25G TCIAC2303-5G
5-氯-2-氟吡啶 ≥96.0% (by GC)
5-氯-2-氟吡啶
分子式: C₅H₃ClFN
分子量: 131.54 g/mol
沸点: 149°C
闪点: 52°C
MDL编号: MFCD04112513
CAS号: 1480-65-5
UN号: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d27eddd9f3c2024bb7fcc23704472196ac85ea39&vid=d27eddd9f3c2024bb7fcc23704472196ac85ea39&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR