3,3,5-Trimethylcaproic acid ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 3,3,5-Trimethylhexanoic acid, 3,5,5-Trimethylhexanoicacid, 3,5,5-Trimethylhexanoic acid

T0630-25ML T0630-500ML
TCIAT0630-25MLEA 0 CNY
TCIAT0630-25ML TCIAT0630-500ML
3,3,5-Trimethylcaproic acid ≥98.0% (by titrimetric analysis)
3,3,5-Trimethylcaproic acid
分子式: C₉H₁₈O₂
沸点: 120°C
闪点: 125°C
MDL编号: MFCD00020507
CAS号: 3302-10-1
EINECS: 221-975-0
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d8d6d07e4af72d1ed4975644f9da3552995fc82&vid=3d8d6d07e4af72d1ed4975644f9da3552995fc82&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR