2,5-二甲基吡嗪 ≥80.0% (by GC) (contains 2,6-isomer)
供应商: TCI
|
Warning
|
D1526-25ML
D1526-500ML
TCIAD1526-25MLEA
0
CNY
TCIAD1526-25ML
TCIAD1526-500ML
2,5-二甲基吡嗪 ≥80.0% (by GC) (contains 2,6-isomer)
2,5-二甲基吡嗪
分子式:
C₆H₈N₂ 分子量: 108.14 g/mol 沸点: 155 溶点: 15 闪点: 64°C 存储条件: Ambient |
MDL编号:
MFCD00006147 CAS号: 123-32-0 EINECS: 204-618-3 |
|
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 80.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...