(3-氯丙基)三甲氧基硅烷 ≥97.0% (by GC)
供应商: TCI
Warning
Synonyms: Dow Corning 产品 Z-6076
C0840-25ML C0840-500ML
TCIAC0840-25MLEA 0 CNY
TCIAC0840-25ML TCIAC0840-500ML
(3-氯丙基)三甲氧基硅烷 ≥97.0% (by GC)
(3-氯丙基)三甲氧基硅烷
分子式: ClH₂CCH₂CH₂Si(OCH₃)₃
沸点: 195°C
溶点: –50 °C
密度: 1,082 g/cm³ (20 °C)
闪点: 78°C
存储条件: Ambient
MDL编号: MFCD00000997
CAS号: 2530-87-2
EINECS: 219-787-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df1c17342d13ea55ec4b3aefb4d92b628575cd8b&vid=df1c17342d13ea55ec4b3aefb4d92b628575cd8b&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %