2-甲氧基苯基硼酸 (contains varying amounts of Anhydride)

供应商: TCI
Warning

Synonyms: 邻甲氧基苯硼酸

M1261-1G M1261-25G M1261-5G
TCIAM1261-1GEA 0 CNY
TCIAM1261-1G TCIAM1261-25G TCIAM1261-5G
2-甲氧基苯基硼酸 (contains varying amounts of Anhydride)
2-甲氧基苯基硼酸
分子式: C₇H₉BO₃
分子量: 151.96 g/mol
溶点: 165
MDL编号: MFCD00236047
CAS号: 5720-06-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f5a52e64c82ad66e2b8ca76ffcbb96fc2a4e491&vid=0f5a52e64c82ad66e2b8ca76ffcbb96fc2a4e491&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 113.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR