1-乙炔基-1-环己醇 ≥98.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
1-Ethynyl-1-cyclohexanol
E0297-25G
E0297-500G
TCIAE0297-25GEA
0
CNY
TCIAE0297-25G
TCIAE0297-500G
1-乙炔基-1-环己醇 ≥98.0% (by GC)
1-乙炔基-1-环己醇
分子式:
C₈H₁₂O 分子量: 124.18 g/mol 沸点: 174°C 存储条件: Refrigerator |
MDL编号:
MFCD00003858 CAS号: 78-27-3 EINECS: 201-100-9 UN号: 2811 ADR: 6.1 III |
|
产品技术规格
Appearance | White to Orange to Green powder to crystaline |
Purity (GC) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...