高氟叔丁醇 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
全氟叔丁醇, 1,1,1,3,3,3-六氟-2-三氟甲基-2-丙醇
N0692-1G
N0692-5G
TCIAN0692-1GEA
0
CNY
TCIAN0692-1G
TCIAN0692-5G
高氟叔丁醇 ≥98.0% (by GC)
高氟叔丁醇
分子式:
C₄HF₉O 沸点: 84 °C (1013 hPa) 溶点: –17 °C 密度: 1,693 g/cm³ (20 °C) |
MDL编号:
MFCD00042092 CAS号: 2378-02-1 EINECS: 219-157-3 ADR: 6.1,II |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...