2,2-二苯基丙腈 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 2,2-Diphenylpropionitrile

D2534-25G D2534-5G
TCIAD2534-25GEA 0 CNY
TCIAD2534-25G TCIAD2534-5G
2,2-二苯基丙腈 ≥98.0% (by GC)
2,2-二苯基丙腈
分子式: (C₆H₅)₂C(CN)CH₃
沸点: 142°C
密度: 1,0671 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00001846
CAS号: 5558-67-8
EINECS: 226-925-1
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa31a013ba440ca4e5e758032ca1817ad4eccc18&vid=fa31a013ba440ca4e5e758032ca1817ad4eccc18&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR