邻苯二甲酸单甲酯 98%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
单甲基邻苯二甲酸酯
374880250
374881000
ACRO374880250EA
0
CNY
ACRO374880250
ACRO374881000
邻苯二甲酸单甲酯 98%
邻苯二甲酸单甲酯
分子式:
C₉H₈O₄ 分子量: 180.16 g/mol 溶点: 81…86 °C 存储条件: Ambient |
MDL编号:
MFCD00002466 CAS号: 4376-18-5 EINECS: 224-476-6 |
产品技术规格
Appearance | White to off-white Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 81 to 86 °C |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...