3′-氨基-4′-氯乙酰苯胺 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

A1559-25G A1559-5G
TCIAA1559-25GEA 0 CNY
TCIAA1559-25G TCIAA1559-5G
3′-氨基-4′-氯乙酰苯胺 ≥98.0% (by titrimetric analysis)
3′-氨基-4′-氯乙酰苯胺
分子式: C₈H₉ClN₂O
分子量: 184.62 g/mol
MDL编号: MFCD02093411
CAS号: 51867-83-5
EINECS: 257-485-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fdae329ec260b2473e7d9a3b0d3f1f9a8aee7ea3&vid=fdae329ec260b2473e7d9a3b0d3f1f9a8aee7ea3&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Gray to Brown powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 170.0 to 174.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR