4-Amino-1-methylpiperidine ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
1-Methylpiperidin-4-amine
A2046-25G
A2046-5G
TCIAA2046-25GEA
0
CNY
TCIAA2046-25G
TCIAA2046-5G
4-Amino-1-methylpiperidine ≥98.0% (by GC, titration analysis)
4-Amino-1-methylpiperidine
分子式:
C₆H₁₄N₂ 分子量: 114.1888 g/mol 沸点: 83°C |
MDL编号:
MFCD01570549 CAS号: 41838-46-4 UN号: 2920 ADR: 8 II |
|
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...