3',4'-Dichlorosalicylanilide ≥98.0% (by total nitrogen basis)

供应商: TCI

D1077-1G D1077-5G
TCIAD1077-1GEA 0 CNY
TCIAD1077-1G TCIAD1077-5G
3',4'-Dichlorosalicylanilide ≥98.0% (by total nitrogen basis)
3',4'-Dichlorosalicylanilide
分子式: C₁₃H₉Cl₂NO₂
MDL编号: MFCD00020023
CAS号: 24448-73-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=432a9e55a77ccd914248b2216dd73e51236d6e68&vid=432a9e55a77ccd914248b2216dd73e51236d6e68&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 212.0 to 217.0 deg-C
Solubility in hot Acetone almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR