Ethyl glycidyl ether ≥98.0% (通过GC)

供应商: TCI
Warning

E0465-25ML E0465-500ML
TCIAE0465-25MLEA 0 CNY
TCIAE0465-25ML TCIAE0465-500ML
Ethyl glycidyl ether ≥98.0% (通过GC)
Ethyl glycidyl ether
分子式: C₅H₁₀O₂
沸点: 128°C
MDL编号: MFCD00085264
CAS号: 4016-11-9
EINECS: 223-671-3
UN号: 2752
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2b6c20437a681a0469d0fb93002c703dc34047e&vid=f2b6c20437a681a0469d0fb93002c703dc34047e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Specific gravity (20/20) 0.9450 to 0.9480
Refractive index n20/D 1.4070 to 1.4090


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR