异丙基缩水甘油醚 ≥96.0% (通过GC)
供应商: TCI
|
Danger
|
G0255-25ML
G0255-500ML
TCIAG0255-25MLEA
0
CNY
TCIAG0255-25ML
TCIAG0255-500ML
异丙基缩水甘油醚 ≥96.0% (通过GC)
异丙基缩水甘油醚
分子式:
C₆H₁₂O₂ 沸点: 132°C 闪点: 33°C |
MDL编号:
MFCD00005139 CAS号: 4016-14-2 EINECS: 223-672-9 UN号: 3271 ADR: 3,III |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...