4-tert-Butylphthalic anhydride ≥98.0% (通过GC,滴定分析)

供应商: TCI
Warning

B1337-25G B1337-5G
TCIAB1337-25GEA 0 CNY
TCIAB1337-25G TCIAB1337-5G
4-tert-Butylphthalic anhydride ≥98.0% (通过GC,滴定分析)
4-tert-Butylphthalic anhydride
分子式: C₁₂H₁₂O₃
沸点: 136°C
MDL编号: MFCD00060127
CAS号: 32703-79-0
EINECS: 251-165-2

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Morpholine Method) min. 98.0 %
Melting point 77.0 to 81.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR