2-Iodo-3-nitrotoluene ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2-Iodo-1-methyl-3-nitrobenzene
I0799-25G
I0799-5G
TCIAI0799-25GEA
0
CNY
TCIAI0799-25G
TCIAI0799-5G
2-Iodo-3-nitrotoluene ≥98.0% (by GC)
2-Iodo-3-nitrotoluene
分子式:
C₇H₆INO₂ 分子量: 263.0300 g/mol |
MDL编号:
MFCD00091040 CAS号: 6277-17-4 |
|
产品技术规格
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 65.0 to 69.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...