3-Bromo-5-methylanisole ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 3-Bromo-5-methoxytoluene, 1-Bromo-3-methoxy-5-methylbenzene

B3526-1G B3526-5G
TCIAB3526-1GEA 0 CNY
TCIAB3526-1G TCIAB3526-5G
3-Bromo-5-methylanisole ≥98.0% (by GC)
3-Bromo-5-methylanisole
分子式: C₈H₉BrO
分子量: 201.0600 g/mol
沸点: 122°C
MDL编号: MFCD08061916
CAS号: 29578-83-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c04e0e93f0d129da695543e7f97c9e31043247f0&vid=c04e0e93f0d129da695543e7f97c9e31043247f0&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Red clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR