3-Bromo-2-fluorotoluene ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 1-Bromo-2-fluoro-3-methylbenzene

B2969-1G B2969-5G
TCIAB2969-1GEA 0 CNY
TCIAB2969-1G TCIAB2969-5G
3-Bromo-2-fluorotoluene ≥98.0% (by GC)
3-Bromo-2-fluorotoluene
分子式: C₇H₆BrF
分子量: 189.02 g/mol
沸点: 186°C
MDL编号: MFCD04972694
CAS号: 59907-12-9
EINECS: 261-981-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d019af9d94753379fddce6554ed5bcc7a421e4e&vid=6d019af9d94753379fddce6554ed5bcc7a421e4e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR