4-甲氧基苯乙酸甲酯 ≥97.0% (by GC)

供应商: TCI
Warning

Synonyms: Methyl 4-methoxyphenylacetate

M1330-25G M1330-500G
TCIAM1330-25GEA 0 CNY
TCIAM1330-25G TCIAM1330-500G
4-甲氧基苯乙酸甲酯 ≥97.0% (by GC)
4-甲氧基苯乙酸甲酯
分子式: C₁₀H₁₂O₃
分子量: 180.23 g/mol
MDL编号: MFCD00008454
CAS号: 23786-14-3
EINECS: 245-886-1
UN号: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3437820cc8b341e1a635015664d3fdb669deb65d&vid=3437820cc8b341e1a635015664d3fdb669deb65d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR