Methyl (3-methoxyphenyl)acetate ≥98.0% (by GC)
供应商: TCI
Synonyms:
Methyl 3-Methoxyphenylacetate
M1331-25G
M1331-5G
TCIAM1331-25GEA
0
CNY
TCIAM1331-25G
TCIAM1331-5G
Methyl (3-methoxyphenyl)acetate ≥98.0% (by GC)
Methyl (3-methoxyphenyl)acetate
分子式:
C₁₀H₁₂O₃ 分子量: 180.23 g/mol |
MDL编号:
MFCD00017205 CAS号: 18927-05-4 |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...