4-(4-Fluorophenoxy)benzonitrile ≥98.0% (by GC)

供应商: TCI

F0532-1G F0532-5G
TCIAF0532-1GEA 0 CNY
TCIAF0532-1G TCIAF0532-5G
4-(4-Fluorophenoxy)benzonitrile ≥98.0% (by GC)
4-(4-Fluorophenoxy)benzonitrile
分子式: C₁₃H₈FNO
分子量: 213.21 g/mol
MDL编号: MFCD01815090
CAS号: 215589-24-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b6f6ba17737f53e45ea0ba8f5c8b836ae4cdb99&vid=4b6f6ba17737f53e45ea0ba8f5c8b836ae4cdb99&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 66.0 to 71.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR