2-乙基-4,5-二甲基噻唑 ≥98.0% (by GC)

供应商: TCI

E0718-25G E0718-5G
TCIAE0718-25GEA 0 CNY
TCIAE0718-25G TCIAE0718-5G
2-乙基-4,5-二甲基噻唑 ≥98.0% (by GC)
2-乙基-4,5-二甲基噻唑
分子式: C₇H₁₁NS
沸点: 185°C
闪点: 74°C
MDL编号: MFCD00053125
CAS号: 873-64-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa254077edccc01ac0f5b65622b902eeb0dfca8a&vid=fa254077edccc01ac0f5b65622b902eeb0dfca8a&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR