2-Ethylthiazole ≥98.0% (通过GC,滴定分析)

供应商: TCI
Warning

E0470-25G E0470-5G
TCIAE0470-25GEA 0 CNY
TCIAE0470-25G TCIAE0470-5G
2-Ethylthiazole ≥98.0% (通过GC,滴定分析)
2-Ethylthiazole
分子式: C₅H₇NS
沸点: 148°C
MDL编号: MFCD00085193
CAS号: 15679-09-1
EINECS: 239-755-8
UN号: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2915d8a67c59638e413f2bd6564b49532baee0d&vid=c2915d8a67c59638e413f2bd6564b49532baee0d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR