3-异丙氧基丙胺 ≥98.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: IPOPA

I0162-25ML I0162-500ML
TCIAI0162-25MLEA 0 CNY
TCIAI0162-25ML TCIAI0162-500ML
3-异丙氧基丙胺 ≥98.0% (by GC, titration analysis)
3-异丙氧基丙胺
分子式: C₆H₁₅NO
沸点: 144°C
闪点: 39°C
MDL编号: MFCD00008220
CAS号: 2906-12-9
EINECS: 220-816-2
UN号: 2734
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef06b8c5858921fd9486e664a931086e46d2461f&vid=ef06b8c5858921fd9486e664a931086e46d2461f&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR