3-Iodo-p-anisidine ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 3-Iodo-4-methoxyaniline

I0716-25G I0716-5G
TCIAI0716-25GEA 0 CNY
TCIAI0716-25G TCIAI0716-5G
3-Iodo-p-anisidine ≥98.0% (by GC, titration analysis)
3-Iodo-p-anisidine
分子式: C₇H₈INO
分子量: 249.0490 g/mol
MDL编号: MFCD06656566
CAS号: 74587-12-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=001ef24b2218bbd2c998ff875092f979f30f4a2e&vid=001ef24b2218bbd2c998ff875092f979f30f4a2e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 72.0 to 75.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR