2,5-二氯-3-甲基噻吩 ≥96.0% (通过GC)
供应商: TCI
|
Danger
|
D3680-1G
D3680-5G
TCIAD3680-1GEA
0
CNY
TCIAD3680-1G
TCIAD3680-5G
2,5-二氯-3-甲基噻吩 ≥96.0% (通过GC)
2,5-二氯-3-甲基噻吩
分子式:
C₅H₄Cl₂S 沸点: 185°C 闪点: 77°C 存储条件: Refrigerator |
MDL编号:
MFCD00022490 CAS号: 17249-90-0 ADR: 6.1,III |
|
产品技术规格
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...