N,N′-二异丙基乙二胺 ≥97.0% (通过GC,滴定分析)
供应商: TCI
|
Danger
|
D3923-25G
D3923-5G
TCIAD3923-25GEA
0
CNY
TCIAD3923-25G
TCIAD3923-5G
N,N′-二异丙基乙二胺 ≥97.0% (通过GC,滴定分析)
N,N′-二异丙基乙二胺
分子式:
C₈H₂₀N₂ 沸点: 170°C 闪点: 52°C |
MDL编号:
MFCD00008864 CAS号: 4013-94-9 EINECS: 223-666-6 UN号: 2733 ADR: 3,III |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...