2-Fluoro-m-toluidine ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
2-Fluoro-3-methylaniline, 3-Amino-2-fluorotoluene
F0554-1G
F0554-5G
TCIAF0554-1GEA
0
CNY
TCIAF0554-1G
TCIAF0554-5G
2-Fluoro-m-toluidine ≥98.0% (by GC, titration analysis)
2-Fluoro-m-toluidine
分子式:
C₇H₈FN 分子量: 125.14 g/mol 沸点: 87°C 存储条件: Refrigerator |
MDL编号:
MFCD06410915 CAS号: 1978-33-2 |
|
产品技术规格
Appearance | Colorless to Light yellow to Red clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...