2-氟-4-甲基苯硼酸 (contains varying amounts of Anhydride)
供应商: TCI
|
Warning
|
F0683-1G
F0683-5G
TCIAF0683-1GEA
0
CNY
TCIAF0683-1G
TCIAF0683-5G
2-氟-4-甲基苯硼酸 (contains varying amounts of Anhydride)
2-氟-4-甲基苯硼酸
分子式:
C₇H₈BFO₂ |
MDL编号:
MFCD05664239 CAS号: 170981-26-7 |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 97.0 to 113.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...